accessibility menu, dialog, popup

UserName

CAS RN 353262-04-1

ONSFFF

CAS RN 353262-04-1

IUPAC Name: 4-{[3-(trifluoromethyl)phenyl]methyl}-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 4-{[3-(trifluoromethyl)phenyl]methyl}-2H-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 323.33
Molecular Formula: C16H12F3NOS
InChi Key: ZUIJXKLTUFCDGO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CN2C(=O)CSC3=CC=CC=C23)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results