accessibility menu, dialog, popup

UserName

CAS RN 3536-96-7

H2CMgCl

CAS RN 3536-96-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 86.8
Molecular Formula: C2H3ClMg
InChi Key: IJMWREDHKRHWQI-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].C=[CH-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)