CAS RN 3555-47-3
CAS RN 3555-47-3
IUPAC Name:
tetratrimethylsilyl silicate
Synonyms:
tetrakis(trimethylsiloxy)silane
Molecular Weight (g/mol):
384.84
Molecular Formula:
C12H36O4Si5
InChi Key:
VNRWTCZXQWOWIG-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C