CAS RN 357-70-0
CAS RN 357-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Galantamine Hydrobromide, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Galanthamine, MedChemExpress
MedChemExpress Galanthamine is a potent acetylcholinesterase (AChE) inhibitor with an IC50 of 500 nM.