accessibility menu, dialog, popup

UserName

HO

CAS RN 3597-91-9

IUPAC Name: {[1,1'-biphenyl]-4-yl}methanol
Synonyms: [1,1'-biphenyl]-4-ylmethanol
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: AXCHZLOJGKSWLV-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results