accessibility menu, dialog, popup

UserName

CAS RN 36099-51-1

CH3O

CAS RN 36099-51-1

IUPAC Name: 3-methyl-7-oxabicyclo[4.1.0]heptane
Synonyms: 3-methyl-7-oxabicyclo[4.1.0]heptane
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: ULPDSNLBZMHGPI-UHFFFAOYNA-N
SMILES: CC1CCC2OC2C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results