accessibility menu, dialog, popup

UserName

CAS RN 36122-35-7

IUPAC Name: 3-phenyl-2,5-dihydrofuran-2,5-dione
Synonyms: 3-phenylfuran-2,5-dione
Molecular Weight (g/mol): 174.16
Molecular Formula: C10H6O3
InChi Key: QZYCWJVSPFQUQC-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)