accessibility menu, dialog, popup

UserName

CAS RN 361393-92-2

CAS RN 361393-92-2

IUPAC Name: 4-(bromomethyl)-2-chloro-1-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-2-chloro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 273.48
Molecular Formula: C8H5BrClF3
InChi Key: DRGNXDDQVUFCEF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results