accessibility menu, dialog, popup

UserName

CAS RN 362661-83-4

NNO

CAS RN 362661-83-4

IUPAC Name: pyrazolo[1,5-a]pyridine-7-carbaldehyde
Synonyms: pyrazolo[1,5-a]pyridine-7-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: SGNBVQKSJWCGOJ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC2=CC=NN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results