accessibility menu, dialog, popup

UserName

CAS RN 363-03-1

OO

CAS RN 363-03-1

IUPAC Name: 2-phenylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2-phenylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 184.19
Molecular Formula: C12H8O2
InChi Key: RLQZIECDMISZHS-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)C(=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results