CAS RN 3633-92-9
CAS RN 3633-92-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desacetyl Vindoline, TRC
CAS: 3633-92-9 Molecular Formula: C23H30N2O5 Molecular Weight (g/mol): 414.49 Synonym: (2β,3β,4β,5α,12R,19α)-6,7-Didehydro-3,4-dihydroxy-16-methoxy-1-methyl-aspidospermidine-3-carboxylic Acid Methyl Ester,4-Desacetylvindoline,Deacetylvindoline,NSC 91993 IUPAC Name: methyl (3aR,3a1R,4R,5S,5aR,10bR)-3a-ethyl-4,5-dihydroxy-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate SMILES: [H][C@]12[C@](C=CCN2CC3)(CC)[C@@H](O)[C@@](C(OC)=O)(O)[C@@]4([H])[C@]13C5=CC=C(OC)C=C5N4C