accessibility menu, dialog, popup

UserName

CAS RN 3662-78-0

CH3OONCS

CAS RN 3662-78-0

IUPAC Name: methyl 4-isothiocyanatobenzoate
Synonyms: methyl 4-isothiocyanatobenzoate
Molecular Weight (g/mol): 193.22
Molecular Formula: C9H7NO2S
InChi Key: WIZODHILGBTPPA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results