accessibility menu, dialog, popup

UserName

NH2NHNFFF

CAS RN 3671-66-7

IUPAC Name: 2-(trifluoromethyl)-1H-1,3-benzodiazol-6-amine
Synonyms: 2-(trifluoromethyl)-3H-1,3-benzodiazol-5-amine
Molecular Weight (g/mol): 201.15
Molecular Formula: C8H6F3N3
InChi Key: CKEKFQLHCAZGSP-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=C(NC2=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)