accessibility menu, dialog, popup

UserName

CAS RN 3687-18-1

H2NSOHOO

CAS RN 3687-18-1

IUPAC Name: 3-aminopropane-1-sulfonic acid
Synonyms: homotaurine 3-aps tramiprosate
Molecular Weight (g/mol): 139.17
Molecular Formula: C3H9NO3S
InChi Key: SNKZJIOFVMKAOJ-UHFFFAOYSA-N
SMILES: NCCCS(O)(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results