accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 36983-31-0

IUPAC Name: ethyl 2,4-dioxoheptanoate
Synonyms: ethyl 2,4-dioxoheptanoate
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: DMAUJRWSYKLVHX-UHFFFAOYSA-N
SMILES: CCCC(=O)CC(=O)C(=O)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results