accessibility menu, dialog, popup

UserName

OHF

CAS RN 371-62-0

IUPAC Name: 2-fluoroethan-1-ol
Synonyms: 2-fluoroethanol
Molecular Weight (g/mol): 64.06
Molecular Formula: C2H5FO
InChi Key: GGDYAKVUZMZKRV-UHFFFAOYSA-N
SMILES: OCCF
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)