accessibility menu, dialog, popup

UserName

CAS RN 37142-46-4

ClOBr

CAS RN 37142-46-4

IUPAC Name: 1-(3-bromopropoxy)-3-chlorobenzene
Synonyms: 1-(3-bromopropoxy)-3-chlorobenzene
Molecular Weight (g/mol): 249.53
Molecular Formula: C9H10BrClO
InChi Key: MVKLHPFBHSJVLA-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(OCCCBr)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)