accessibility menu, dialog, popup

UserName

CAS RN 371935-79-4

ONNNONOHHCl

CAS RN 371935-79-4

IUPAC Name: 3-[6-(morpholin-4-yl)-8-oxa-3,5,10-triazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl]phenol hydrochloride
Synonyms: 3-[6-(morpholin-4-yl)-8-oxa-3,5,10-triazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl]phenol hydrochloride
Molecular Weight (g/mol): 384.82
Molecular Formula: C19H17ClN4O3
InChi Key: XSQMYBFFYPTMFE-UHFFFAOYSA-N
SMILES: Cl.OC1=CC=CC(=C1)C1=NC2=C(OC3=C2C=CC=N3)C(=N1)N1CCOCC1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results