accessibility menu, dialog, popup

UserName

CAS RN 37488-45-2

HNSNH2

CAS RN 37488-45-2

IUPAC Name: 1H-pyrrole-2-carbothioamide
Synonyms: 1H-pyrrole-2-carbothioamide
Molecular Weight (g/mol): 126.18
Molecular Formula: C5H6N2S
InChi Key: KJEMJZMJDZLKRH-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results