accessibility menu, dialog, popup

UserName

CAS RN 375-02-0

FFFFFFFO

CAS RN 375-02-0

IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanal
Synonyms: 2,2,3,3,4,4,4-heptafluorobutanal
Molecular Weight (g/mol): 198.04
Molecular Formula: C4HF7O
InChi Key: IQJZGNJYXIIMGP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)