CAS RN 3759-92-0
CAS RN 3759-92-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Furaltadone hydrochloride, MedChemExpress
MedChemExpress Furaltadone hydrochloride, a nitrofuran drug, has the potential for the study in infections of chickens with salmonella enteritidis. Furaltadone is inhibitory and bactericidal in vitro for staphylococci .