accessibility menu, dialog, popup

UserName

CAS RN 3761-42-0

IUPAC Name: O-4-methanesulfonyl-3-methylphenyl O,O-dimethyl phosphorothioate
Synonyms: O-4-methanesulfonyl-3-methylphenyl O,O-dimethyl phosphorothioate
Molecular Weight (g/mol): 310.32
Molecular Formula: C10H15O5PS2
InChi Key: ZDHYERRNXRANLI-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=CC=C(C(C)=C1)S(C)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results