CAS RN 37853-61-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tetrabromobisphenol A Dimethyl Ether, TRC
CAS: 37853-61-5 Molecular Formula: C17 H16 Br4 O2 Molecular Weight (g/mol): 571.92 Synonym: Tetrabromobisphenol A-dimethyl Ether,Benzene, 1,1'-(1-methylethylidene)bis[3,5-dibromo-4-methoxy-,1,1'-(1-Methylethylidene)bis[3,5-dibromo-4-methoxybenzene],2,2-Bis(3,5-dibromo-4-methoxyphenyl)propane,Tetrabromobisphenol A dimethyl ether,Tetrabromobisphenol A methyl ether IUPAC Name: 1,3-dibromo-5-[2-(3,5-dibromo-4-methoxyphenyl)propan-2-yl]-2-methoxybenzene SMILES: COc1c(Br)cc(cc1Br)C(C)(C)c2cc(Br)c(OC)c(Br)c2