CAS RN 37927-29-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Prednienic Acid, TRC
CAS: 37927-29-0 Molecular Formula: C20 H26 O5 Molecular Weight (g/mol): 346.42 Synonym: Prednienic Acid,Prednisolone Impurity B, (11β,17α)-11,17-Dihydroxy-3-oxo-androsta-1,4-diene-17-carboxylic Acid; IUPAC Name: (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-17-carboxylic acid SMILES: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)O
2
Prednienic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.