accessibility menu, dialog, popup

UserName

CAS RN 380430-68-2

CH3H3CCH3OOHNBOHHO

CAS RN 380430-68-2

IUPAC Name: (3-{[(tert-butoxy)carbonyl]amino}phenyl)boronic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]phenylboronic acid
Molecular Weight (g/mol): 237.06
Molecular Formula: C11H16BNO4
InChi Key: CWLNHPXWZRALFS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results