accessibility menu, dialog, popup

UserName

CH3CH3S

CAS RN 38360-81-5

IUPAC Name: (3,5-dimethylphenyl)sulfanide
Synonyms: (3,5-dimethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: CESBAYSBPMVAEI-UHFFFAOYSA-M
SMILES: CC1=CC([S-])=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results