accessibility menu, dialog, popup

UserName

CAS RN 3839-22-3

OON

CAS RN 3839-22-3

IUPAC Name: 2-cyanobenzoate
Synonyms: 2-cyanobenzoate
Molecular Weight (g/mol): 146.13
Molecular Formula: C8H4NO2
InChi Key: DTNSDCJFTHMDAK-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)