CAS RN 38455-90-2
CAS RN 38455-90-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-Methylephedrine Hydrochloride, TRC
CAS: 38455-90-2 Molecular Formula: C11H17NOClH Molecular Weight (g/mol): 215.7 Synonym: (αR)-α-[(1S)-1-(Dimethylamino)ethyl]benzenemethanol hydrochloride (1:1),[R-(R*,S*)]-α-[1-(Dimethylamino)ethyl]benzenemethanol hydrochloride,(-)-Methylephedrine hydrochloride,L-erythro-Methylephedrine hydrochloride,Methy F,N-Methylephedrine hydrochloride,NSC 42289,L-N-Methylephedrine hydrochloride IUPAC Name: (1R,2S)-2-(dimethylamino)-1-phenylpropan-1-ol;hydrochloride SMILES: Cl.C[C@@H]([C@H](O)c1ccccc1)N(C)C