accessibility menu, dialog, popup

UserName

CAS RN 38582-18-2

ZnOOOO

CAS RN 38582-18-2

IUPAC Name: zinc(2+) bis(4-cyclohexylbutanoate)
Synonyms: zinc(2+) bis(4-cyclohexylbutanoate)
Molecular Weight (g/mol): 403.87
Molecular Formula: C20H34O4Zn
InChi Key: KVSXXOUZYJZCPY-UHFFFAOYSA-L
SMILES: [Zn++].[O-]C(=O)CCCC1CCCCC1.[O-]C(=O)CCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results