CAS RN 3862-73-5
IUPAC Name:
2,3,4-trifluoroaniline
Synonyms:
benzenamine, 2,3,4-trifluoro-
Molecular Weight (g/mol):
147.1
Molecular Formula:
C6H4F3N
InChi Key:
WRDGNXCXTDDYBZ-UHFFFAOYSA-N
SMILES:
NC1=CC=C(F)C(F)=C1F
1
–
1
of
1
results
1
2,3,4-Trifluoroaniline, 98%
CAS: 3862-73-5 Molecular Formula: C6H4F3N Molecular Weight (g/mol): 147.1 MDL Number: MFCD00011737 InChI Key: WRDGNXCXTDDYBZ-UHFFFAOYSA-N Synonym: 2,3,4-trifluorobenzenamine,benzenamine, 2,3,4-trifluoro,2,3,4-trifluoraniline,2,3,4-trifluoro-phenylamine,aniline, 2,3,4-trifluoro,2,3,4-trifluoro aniline,2,3,4-trifluorophenylamine,pubchem2297,acmc-1clgf,2.3.4-trifluoroaniline PubChem CID: 77468 IUPAC Name: 2,3,4-trifluoroaniline SMILES: C1=CC(=C(C(=C1N)F)F)F