accessibility menu, dialog, popup

UserName

CAS RN 38701-74-5

OFFFFFF2H2H

CAS RN 38701-74-5

IUPAC Name: hexafluoro(²H)propan-2-(²H)ol
Synonyms: hexafluoro(²H)propan-2-(²H)ol
Molecular Weight (g/mol): 170.05
Molecular Formula: C3H2F6O
InChi Key: BYEAHWXPCBROCE-AWPANEGFSA-N
SMILES: [2H]OC([2H])(C(F)(F)F)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)