accessibility menu, dialog, popup

UserName

CAS RN 38721-52-7

BH3CCH3H3CCH3CH3H3CHLi

CAS RN 38721-52-7

IUPAC Name: boron(3+) lithium(1+) tris(butan-2-ide) hydride
Synonyms: L-selectride
Molecular Weight (g/mol): 190.11
Molecular Formula: C12H28BLi
InChi Key: NUYXPUUFMRFXCC-UHFFFAOYSA-N
SMILES: [H-].[Li+].[B+3].CC[CH-]C.CC[CH-]C.CC[CH-]C
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (7)
  • (2)
  • (2)
  • (2)
  • (1)