accessibility menu, dialog, popup

UserName

CAS RN 39177-22-5

IUPAC Name: 3-fluoro-4-phenoxyaniline
Synonyms: 3-fluoro-4-phenoxyaniline
Molecular Weight (g/mol): 203.22
Molecular Formula: C12H10FNO
InChi Key: SYUZQKSUMLAOIB-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(OC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results