accessibility menu, dialog, popup

UserName

CAS RN 39191-07-6

CH3HNCl

CAS RN 39191-07-6

IUPAC Name: [(3-chlorophenyl)methyl](methyl)amine
Synonyms: [(3-chlorophenyl)methyl](methyl)amine
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: ZPNLAQVYPIAHTO-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results