accessibility menu, dialog, popup

UserName

CAS RN 39232-91-2

HClCH3OHNNH2

CAS RN 39232-91-2

IUPAC Name: hydrogen (3-methoxyphenyl)hydrazine chloride
Synonyms: hydrogen (3-methoxyphenyl)hydrazine chloride
Molecular Weight (g/mol): 174.63
Molecular Formula: C7H11ClN2O
InChi Key: GMXFZBZOVZOYNQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=CC(NN)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results