accessibility menu, dialog, popup

UserName

CH3OOFNOO

CAS RN 393-85-1

IUPAC Name: methyl 5-fluoro-2-nitrobenzoate
Synonyms: methyl 5-fluoro-2-nitrobenzoate
Molecular Weight (g/mol): 199.14
Molecular Formula: C8H6FNO4
InChi Key: LCAUVFVOLNRVKG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CC(F)=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results