accessibility menu, dialog, popup

UserName

OOFNOO

CAS RN 394-01-4

IUPAC Name: 4-fluoro-2-nitrobenzoate
Synonyms: 4-fluoro-2-nitrobenzoate
Molecular Weight (g/mol): 184.1
Molecular Formula: C7H3FNO4
InChi Key: YLUCXHMYRQUERW-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(F)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results