accessibility menu, dialog, popup

UserName

CH3OHCH3CH3H3C

CAS RN 3970-62-5

IUPAC Name: 2,2-dimethylpentan-3-ol
Synonyms: 2,2-dimethyl-3-pentanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: HMSVXZJWPVIVIV-UHFFFAOYNA-N
SMILES: CCC(O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)