CAS RN 398507-55-6
CAS RN 398507-55-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lodenafil Carbonate, TRC
CAS: 398507-55-6 Chemical Name or Material: Lodenafil Carbonate InChI Formula: InChI=1S/C47H62N12O11S2/c1-7-11-35-39-41(54(5)52-35)45(60)50-43(48-39)33-29-31(13-15-37(33)67-9-3)71(63,64)58-21-17-56(18-22-58)25-27-69-47(62)70-28-26-57-19-23-59(24-20-57)72(65,66)32-14-16-38(68-10-4)34(30-32)44-49-40-36(12-8-2)53-55(6)42(40)46(61)51-44/h13-16,29-30H,7-12,17-28H2,1-6H3,(H,48,50,60)(H,49,51,61) IUPAC Name: bis[2-[4-[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]sulfonylpiperazin-1-yl]ethyl] carbonate Molecular Formula: C47 H62 N12 O11 S2 Molecular Weight (g/mol): 1035.2