CAS RN 39856-58-1
CAS RN 39856-58-1
IUPAC Name:
2-bromopyridin-3-amine
Synonyms:
2-bromopyridin-3-amine
Molecular Weight (g/mol):
173.01
Molecular Formula:
C5H5BrN2
InChi Key:
HKDVVTLISGIPFE-UHFFFAOYSA-N
SMILES:
NC1=CC=CN=C1Br
1
–
2
of
2
results
1
3-Amino-2-bromopyridine, 97%
CAS: 39856-58-1 Molecular Formula: C5H5BrN2 Molecular Weight (g/mol): 173.013 MDL Number: MFCD00234064 InChI Key: HKDVVTLISGIPFE-UHFFFAOYSA-N Synonym: 3-amino-2-bromopyridine,2-bromo-3-aminopyridine,2-bromo-pyridineamine,3-pyridinamine, 2-bromo,2-bromo-3-pyridinamine,2-bromo-3-pyridylamine,2-bromopyridine-3-amine,3-amino-2-bromo-pyridine,2-bromo-3-pyridineamine,2-bromo-pyridin-3-ylamine PubChem CID: 642816 IUPAC Name: 2-bromopyridin-3-amine SMILES: C1=CC(=C(N=C1)Br)N