accessibility menu, dialog, popup

UserName

NH2ClFFF

CAS RN 39885-50-2

IUPAC Name: 2-chloro-4-(trifluoromethyl)aniline
Synonyms: 2-chloro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: MBBUTABXEITVNY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Cl)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results