accessibility menu, dialog, popup

UserName

NClClFFF

CAS RN 39890-98-7

IUPAC Name: 2,6-dichloro-4-(trifluoromethyl)pyridine
Synonyms: 2,6-dichloro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.98
Molecular Formula: C6H2Cl2F3N
InChi Key: KVNQWVYYVLCZKK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=NC(Cl)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)