CAS RN 39943-37-8
CAS RN 39943-37-8
IUPAC Name:
1-[(dimethylamino)methyl]cyclopentane-1-carbaldehyde
Synonyms:
1-[(dimethylamino)methyl]cyclopentane-1-carbaldehyde
Molecular Weight (g/mol):
155.24
Molecular Formula:
C9H17NO
InChi Key:
NEHVBXDPRDUMRT-UHFFFAOYSA-N
SMILES:
CN(C)CC1(CCCC1)C=O
1
–
1
of
1
results
1
1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde, 97%, Thermo Scientific™
CAS: 39943-37-8 Molecular Formula: C9H17NO Molecular Weight (g/mol): 155.24 MDL Number: MFCD08064242 InChI Key: NEHVBXDPRDUMRT-UHFFFAOYSA-N Synonym: 1-dimethylaminomethyl cyclopentanecarboxaldehyde,cyclopentanecarboxaldehyde, 1-dimethylamino methyl-9ci,1-dimethylamino methyl cyclopentane-1-carbaldehyde,acmc-20alc3,1-dimethylaminomethylcyclopentancarboxaldehyd,1-dimethylamino methyl cyclopentanecarbaldehyde PubChem CID: 12610330 IUPAC Name: 1-[(dimethylamino)methyl]cyclopentane-1-carbaldehyde SMILES: CN(C)CC1(CCCC1)C=O