accessibility menu, dialog, popup

UserName

HOBOHNF

CAS RN 401815-98-3

IUPAC Name: (2-fluoropyridin-4-yl)boronic acid
Synonyms: 2-fluoropyridin-4-ylboronic acid
Molecular Weight (g/mol): 140.91
Molecular Formula: C5H5BFNO2
InChi Key: WXGBZJJAGLSBPR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=NC(F)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)