accessibility menu, dialog, popup

UserName

CAS RN 40248-84-8

SHHO

CAS RN 40248-84-8

IUPAC Name: 3-sulfanylphenol
Synonyms: 3-hydroxythiophenol
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: DOFIAZGYBIBEGI-UHFFFAOYSA-N
SMILES: OC1=CC(S)=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results