accessibility menu, dialog, popup

UserName

CAS RN 40499-83-0

OHHN

CAS RN 40499-83-0

IUPAC Name: pyrrolidin-3-ol
Synonyms: pyrrolidin-3-ol
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: JHHZLHWJQPUNKB-UHFFFAOYNA-N
SMILES: OC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results