CAS RN 4050-45-7
CAS RN 4050-45-7
IUPAC Name:
(2E)-hex-2-ene
Synonyms:
2-hexene (trans) 4-octene (trans) hexene - mixture of isomers
Molecular Weight (g/mol):
84.16
Molecular Formula:
C6H12
InChi Key:
RYPKRALMXUUNKS-HWKANZROSA-N
SMILES:
CCC\C=C\C
1
–
3
of
3
results
1
trans-2-Hexene, 99%
CAS: 4050-45-7 Molecular Formula: C6H12 Molecular Weight (g/mol): 84.16 MDL Number: MFCD00009473 InChI Key: RYPKRALMXUUNKS-HWKANZROSA-N Synonym: trans-2-hexene,e-2-hexene,2-hexene, e,hexene, isomer,hexene, isomers,unii-2tz30ggg1a,2e-hexene,e-hex-2-ene,2tz30ggg1a,2-hexene, 2e PubChem CID: 639661 SMILES: CCC\C=C\C
2
trans-2-Hexene, 98+%
CAS: 4050-45-7 Molecular Formula: C6H12 Molecular Weight (g/mol): 84.16 MDL Number: MFCD00009473 InChI Key: RYPKRALMXUUNKS-HWKANZROSA-N Synonym: trans-2-hexene,e-2-hexene,2-hexene, e,hexene, isomer,hexene, isomers,unii-2tz30ggg1a,2e-hexene,e-hex-2-ene,2tz30ggg1a,2-hexene, 2e PubChem CID: 639661 IUPAC Name: (E)-hex-2-ene SMILES: CCC\C=C\C