accessibility menu, dialog, popup

UserName

CAS RN 4052-92-0

ClOSClOO

CAS RN 4052-92-0

IUPAC Name: 3-(chlorosulfonyl)benzoyl chloride
Synonyms: 3-(chlorosulfonyl)benzoyl chloride
Molecular Weight (g/mol): 239.07
Molecular Formula: C7H4Cl2O3S
InChi Key: XWEBTVZIZWEJOO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O

 No Refinements