accessibility menu, dialog, popup

UserName

CAS RN 40957-83-3

H3COOOHOOH

CAS RN 40957-83-3

IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one
Synonyms: glycitein
Molecular Weight (g/mol): 284.27
Molecular Formula: C16H12O5
InChi Key: DXYUAIFZCFRPTH-UHFFFAOYSA-N
SMILES: COC1=C(O)C=C2OC=C(C(=O)C2=C1)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)