CAS RN 41012-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-Fructose 1,6-Bisphosphate Sodium Salt Hydrate, TRC
CAS: 41012-14-0 Molecular Weight (g/mol): 380.11 Synonym: D-Fructose, 1,6-bis(dihydrogen phosphate), sodium salt, hydrate (9CI),D-Fructose 1,6-bisphosphate sodium salt hydrate IUPAC Name: sodium;[(3S,4S,5R)-2,3,4-trihydroxy-5-(phosphonooxymethyl)oxolan-2-yl]methyl hydrogen phosphate;hydrate SMILES: O.[Na+].O[C@H]1[C@H](O)C(O)(COP(=O)(O)[O-])O[C@@H]1COP(=O)(O)O